Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLF11
Entrez Gene ID
8462
Gene Name
Kruppel like factor 11
Gene Aliases
FKLF, FKLF1, MODY7, TIEG2, Tieg3
Chromosome Location
Chr.2: 10043555 - 10054836 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177718.1 NP_001171189.1 1 - 2 203 100
GenBank mRNA AK302880.1 - 1 - 2 168 100
BG701929.1 - 1 - 2 210 100

View Full Product Details