The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261392.1 | NP_001248321.1 | 7 - 8 | 636 | 93 |
NM_001261394.1 | NP_001248323.1 | 8 - 9 | 714 | 93 | |
NM_024894.3 | NP_079170.2 | 8 - 9 | 714 | 93 | |
NR_048552.1 | 7 - 8 | 668 | 93 | ||
XM_011510398.2 | XP_011508700.1 | 7 - 8 | 581 | 93 | |
XM_011510399.2 | XP_011508701.1 | 6 - 7 | 519 | 93 | |
XM_011510400.2 | XP_011508702.1 | 8 - 9 | 702 | 93 | |
XM_011510402.2 | XP_011508704.1 | 8 - 9 | 702 | 93 | |
XM_017004990.1 | XP_016860479.1 | 6 - 7 | 456 | 93 | |
GenBank mRNA | AK024000.1 | - | 7 - 8 | 555 | 93 |
AK024137.1 | - | 8 - 9 | 699 | 93 | |
AK290680.1 | - | 8 - 9 | 678 | 93 | |
AK297169.1 | - | 7 - 8 | 624 | 93 | |
AK299617.1 | - | 7 - 8 | 656 | 93 | |
BC005125.2 | - | 8 - 9 | 694 | 93 | |
BG390065.1 | - | 3 - 4 | 168 | 93 |