The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004464.3 | NP_004455.2 | 1 - 2 | 597 | 79 |
GenBank mRNA | AF535149.1 | - | 1 - 2 | 522 | 79 |
AK291962.1 | - | 1 - 2 | 522 | 79 | |
BC074858.2 | - | 1 - 2 | 409 | 79 | |
BC074859.2 | - | 1 - 2 | 409 | 79 | |
BC131502.1 | - | 1 - 2 | 498 | 79 | |
M37825.1 | - | 1 - 2 | 499 | 79 |