Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIST3H2A
Entrez Gene ID
92815
Gene Name
histone cluster 3, H2a
Gene Aliases
-
Chromosome Location
Chr.1: 228457364 - 228457859 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC001193.1 - 1 - 1 722 60
BC082269.1 - 1 - 1 712 60

View Full Product Details