Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZCCHC18
Entrez Gene ID
644353
Gene Name
zinc finger CCHC-type containing 18
Gene Aliases
PNMA7B, SIZN2
Chromosome Location
Chr.X: 104112416 - 104121330 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143978.2 NP_001137450.1 3 - 3 1335 66
XM_011531012.2 XP_011529314.1 3 - 3 1472 66

View Full Product Details