The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022436.2 | NP_071881.1 | 3 - 4 | 543 | 99 |
XM_005264480.3 | XP_005264537.1 | 3 - 4 | 564 | 99 | |
XM_006712073.3 | XP_006712136.1 | 3 - 4 | 564 | 99 | |
XM_006712074.3 | XP_006712137.1 | 3 - 4 | 564 | 99 | |
XM_011533024.2 | XP_011531326.1 | 3 - 4 | 564 | 99 | |
XM_011533025.2 | XP_011531327.1 | 3 - 4 | 988 | 99 | |
XM_011533026.2 | XP_011531328.1 | 3 - 4 | 564 | 99 | |
GenBank mRNA | AF312715.2 | - | 3 - 4 | 543 | 99 |
AF320293.1 | - | 3 - 4 | 509 | 99 | |
AK091997.1 | - | 2 - 3 | 1170 | 99 | |
BC111541.1 | - | 2 - 3 | 1055 | 99 |