The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030582.3 | NP_085059.2 | 5 - 6 | 1492 | 95 |
NM_130444.2 | NP_569711.2 | 5 - 6 | 2197 | 95 | |
NM_130445.3 | NP_569712.2 | 6 - 7 | 1025 | 95 | |
GenBank mRNA | AF018081.1 | - | 5 - 6 | 1511 | 95 |
AF018082.1 | - | 6 - 7 | 990 | 95 | |
BC063833.1 | - | 6 - 7 | 966 | 95 |