The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181621.3 | NP_853652.1 | 1 - 1 | 666 | 75 |
GenBank mRNA | AB096938.1 | - | 1 - 1 | 656 | 75 |
BX104478.1 | - | 1 - 1 | 683 | 75 | |
W72002.1 | - | 1 - 1 | 75 |