Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C6orf58
Entrez Gene ID
352999
Gene Name
chromosome 6 open reading frame 58
Gene Aliases
LEG1
Chromosome Location
Chr.6: 127577174 - 127591818 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001010905.2 NP_001010905.1 4 - 5 678 84
GenBank mRNA AK303850.1 - 4 - 5 678 84
BC062712.1 - 4 - 5 682 84

View Full Product Details