The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001161683.1 | NP_001155155.1 | 12 - 13 | 1660 | 62 |
NM_144672.3 | NP_653273.3 | 16 - 17 | 1804 | 62 | |
NM_170664.2 | NP_733764.1 | 7 - 8 | 898 | 62 | |
XM_011545747.2 | XP_011544049.1 | 17 - 18 | 2183 | 62 | |
XM_011545748.2 | XP_011544050.2 | 6 - 7 | 1177 | 62 | |
XM_017022950.1 | XP_016878439.1 | 17 - 18 | 1907 | 62 | |
XM_017022951.1 | XP_016878440.1 | 2 - 3 | 201 | 62 | |
GenBank mRNA | AK093062.1 | - | 7 - 8 | 898 | 62 |
AK125840.1 | - | 12 - 13 | 1660 | 62 | |
AK302733.1 | - | 6 - 7 | 960 | 62 | |
BC129992.1 | - | 16 - 17 | 1801 | 62 | |
BC129993.1 | - | 16 - 17 | 1808 | 62 |