Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CUEDC2
Entrez Gene ID
79004
Gene Name
CUE domain containing 2
Gene Aliases
C10orf66, bA18I14.5
Chromosome Location
Chr.10: 102423245 - 102432666 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024040.2 NP_076945.2 4 - 5 425 73
GenBank mRNA AB025425.1 - 3 - 4 274 73
AB232358.1 - 3 - 4 274 73
BC000262.1 - 4 - 5 355 73
BM756951.1 - 4 - 5 425 73
CX165866.1 - 4 - 5 309 73

View Full Product Details