The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172435.1 | NP_001165906.1 | 13 - 14 | 1280 | 164 |
NM_012233.2 | NP_036365.1 | 13 - 14 | 1280 | 164 | |
XM_011510823.2 | XP_011509125.1 | 13 - 14 | 1280 | 164 | |
XM_011510825.2 | XP_011509127.1 | 13 - 14 | 1280 | 164 | |
GenBank mRNA | AB449877.1 | - | 13 - 14 | 1237 | 164 |
AK022494.1 | - | 13 - 14 | 1252 | 164 | |
BC146809.1 | - | 15 - 16 | 1517 | 164 | |
D31886.1 | - | 13 - 14 | 1239 | 164 |