The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172435.1 | NP_001165906.1 | 8 - 9 | 787 | 114 |
NM_012233.2 | NP_036365.1 | 8 - 9 | 787 | 114 | |
XM_011510823.2 | XP_011509125.1 | 8 - 9 | 787 | 114 | |
XM_011510825.2 | XP_011509127.1 | 8 - 9 | 787 | 114 | |
GenBank mRNA | AB449877.1 | - | 8 - 9 | 744 | 114 |
AK022494.1 | - | 8 - 9 | 759 | 114 | |
BC146809.1 | - | 10 - 11 | 1024 | 114 | |
D31886.1 | - | 8 - 9 | 746 | 114 |