Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFFO1
Entrez Gene ID
25900
Gene Name
intermediate filament family orphan 1
Gene Aliases
HOM-TES-103, IFFO
Chromosome Location
Chr.12: 6539528 - 6556102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_036467.1 5 - 6 1232 143
XM_006718976.3 XP_006719039.1 4 - 5 1145 143
XM_006718977.3 XP_006719040.1 4 - 5 1145 143
XM_011520949.2 XP_011519251.1 4 - 5 1145 143
XM_017019173.1 XP_016874662.1 5 - 6 454 143
GenBank mRNA AK095106.1 - 1 - 2 713 143
BC001790.1 - 5 - 6 1212 143
BC010431.1 - 5 - 6 1212 143
BC023664.2 - 5 - 6 1212 143

View Full Product Details