The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001307977.1 | NP_001294906.1 | 1 - 2 | 148 | 55 |
NM_018093.3 | NP_060563.2 | 2 - 3 | 227 | 55 | |
XM_005274055.2 | XP_005274112.1 | 2 - 3 | 2088 | 55 | |
GenBank mRNA | AK001301.1 | - | 2 - 3 | 227 | 55 |
AK292330.1 | - | 2 - 3 | 597 | 55 | |
AK303186.1 | - | 1 - 2 | 69 | 55 | |
AY500828.1 | - | 1 - 2 | 79 | 55 | |
BC006351.2 | - | 1 - 2 | 70 | 55 |