Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERP44
Entrez Gene ID
23071
Gene Name
endoplasmic reticulum protein 44
Gene Aliases
PDIA10, TXNDC4
Chromosome Location
Chr.9: 99979179 - 100099052 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015051.2 NP_055866.1 4 - 5 497 73
GenBank mRNA AB011145.1 - 4 - 5 421 73
AJ344330.2 - 4 - 5 408 73
AK075024.1 - 4 - 5 485 73
AY359048.1 - 4 - 5 421 73
BC005374.1 - 4 - 5 458 73
BQ881761.1 - 4 - 5 497 73

View Full Product Details