The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017839.4 | NP_060309.2 | 10 - 11 | 1244 | 103 |
XM_011523169.2 | XP_011521471.1 | 10 - 11 | 1144 | 103 | |
XM_017023376.1 | XP_016878865.1 | 11 - 12 | 1218 | 103 | |
GenBank mRNA | AB244718.1 | - | 10 - 11 | 1063 | 103 |
AK000488.1 | - | 10 - 11 | 1171 | 103 | |
AK308386.1 | - | 10 - 11 | 1172 | 103 | |
BC002472.2 | - | 10 - 11 | 1203 | 103 | |
BX641069.1 | - | 5 - 6 | 1146 | 103 |