The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012435.2 | NP_036567.2 | 11 - 12 | 1620 | 89 |
XM_011527895.2 | XP_011526197.1 | 10 - 11 | 1500 | 89 | |
GenBank mRNA | AB001451.1 | - | 11 - 12 | 1494 | 89 |
AL360254.1 | - | 9 - 10 | 1050 | 89 | |
BC048290.1 | - | 1 - 2 | 322 | 89 | |
BY796046.2 | - | 2 - 3 | 437 | 89 | |
CF457169.1 | - | 2 - 3 | 422 | 89 |