Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- UCHL5
- Entrez Gene ID
-
51377
- Gene Name
- ubiquitin C-terminal hydrolase L5
- Gene Aliases
-
CGI-70, INO80R, UCH-L5, UCH37
- Chromosome Location
-
Chr.1: 193012366 - 193060140 on Build GRCh38
View Full Product Details