The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000803.4 | NP_000794.3 | 3 - 4 | 514 | 78 |
NM_001113534.1 | NP_001107006.1 | 3 - 4 | 508 | 78 | |
NM_001113535.1 | NP_001107007.1 | 3 - 4 | 502 | 78 | |
NM_001113536.1 | NP_001107008.1 | 3 - 4 | 496 | 78 | |
XM_005273856.3 | XP_005273913.1 | 3 - 4 | 512 | 78 | |
GenBank mRNA | AK222539.1 | - | 3 - 4 | 505 | 78 |
AK222643.1 | - | 3 - 4 | 496 | 78 | |
BC027894.1 | - | 3 - 4 | 483 | 78 | |
BC058036.1 | - | 3 - 4 | 498 | 78 | |
BC115366.1 | - | 2 - 3 | 284 | 78 | |
BM691283.1 | - | 3 - 4 | 454 | 78 | |
BM849995.1 | - | 3 - 4 | 514 | 78 | |
BY797003.2 | - | 3 - 4 | 477 | 78 | |
CD014063.1 | - | 2 - 3 | 365 | 78 | |
CD692073.1 | - | 3 - 4 | 512 | 78 |