The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194318.3 | NP_919299.3 | 14 - 15 | 1439 | 85 |
XM_006719768.3 | XP_006719831.1 | 15 - 16 | 1629 | 85 | |
XM_011534936.1 | XP_011533238.1 | 13 - 14 | 1308 | 85 | |
XM_011534938.2 | XP_011533240.1 | 13 - 14 | 1277 | 85 | |
XM_017020395.1 | XP_016875884.1 | 15 - 16 | 1532 | 85 | |
GenBank mRNA | AB101481.1 | - | 14 - 15 | 1329 | 85 |
AK291273.1 | - | 14 - 15 | 1428 | 85 | |
AY190526.1 | - | 14 - 15 | 1329 | 85 | |
BC068595.1 | - | 14 - 15 | 1419 | 85 | |
BX647932.1 | - | 1 - 2 | 505 | 85 |