Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SLC38A4
- Entrez Gene ID
-
55089
- Gene Name
- solute carrier family 38 member 4
- Gene Aliases
-
ATA3, NAT3, PAAT, SNAT4
- Chromosome Location
-
Chr.12: 46764761 - 46832422 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK001053.1 |
- |
1 - 2 |
80 |
123 |
View Full Product Details