The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001143824.1 | NP_001137296.1 | 8 - 9 | 861 | 112 |
NM_018018.4 | NP_060488.2 | 9 - 10 | 1053 | 112 | |
XM_005268997.2 | XP_005269054.1 | 8 - 9 | 862 | 112 | |
GenBank mRNA | AB055003.1 | - | 7 - 8 | 685 | 112 |
AF193836.1 | - | 8 - 9 | 841 | 112 | |
AF305814.1 | - | 9 - 10 | 1018 | 112 | |
AK001053.1 | - | 9 - 10 | 929 | 112 | |
AK291168.1 | - | 8 - 9 | 862 | 112 | |
BC069819.1 | - | 8 - 9 | 914 | 112 | |
BC101827.1 | - | 7 - 8 | 698 | 112 | |
BC104913.1 | - | 7 - 8 | 697 | 112 |