Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HDAC7
Entrez Gene ID
51564
Gene Name
histone deacetylase 7
Gene Aliases
HD7, HD7A, HDAC7A
Chromosome Location
Chr.12: 47782710 - 47820612 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098416.3 NP_001091886.1 14 - 15 2019 69
NM_001308090.1 NP_001295019.1 14 - 15 2079 69
NM_015401.4 NP_056216.2 15 - 16 2130 69
XM_011538478.1 XP_011536780.1 15 - 16 2082 69
XM_011538479.1 XP_011536781.1 16 - 17 2524 69
XM_011538480.1 XP_011536782.1 14 - 15 1971 69
XM_011538481.1 XP_011536783.1 15 - 16 1975 69
XM_011538482.1 XP_011536784.1 15 - 16 2059 69
XM_017019455.1 XP_016874944.1 14 - 15 2031 69
GenBank mRNA AK001190.1 - 7 - 8 69
AK024469.1 - 10 - 11 2110 69
AK122588.1 - 2 - 3 1750 69
AK128383.1 - 7 - 8 973 69
AK299292.1 - 15 - 16 2045 69
AK301545.1 - 15 - 16 69
AK303481.1 - 14 - 15 1985 69
AL117455.1 - 11 - 12 1597 69
AY302468.1 - 15 - 16 1935 69
AY321367.1 - 14 - 15 1904 69
BC006453.1 - 4 - 5 554 69
BC020505.2 - 7 - 8 1097 69
BC064840.1 - 14 - 15 1913 69
BT009771.1 - 5 - 6 804 69

View Full Product Details