The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281765.2 | NP_001268694.1 | 10 - 11 | 2683 | 116 |
NM_001281766.2 | NP_001268695.1 | 9 - 10 | 2617 | 116 | |
NM_001281767.2 | NP_001268696.1 | 10 - 11 | 2683 | 116 | |
NM_001318761.1 | NP_001305690.1 | 10 - 11 | 1801 | 116 | |
NM_004439.7 | NP_004430.4 | 10 - 11 | 2680 | 116 | |
NM_182472.4 | NP_872272.2 | 9 - 10 | 2614 | 116 | |
XM_005265653.3 | XP_005265710.1 | 8 - 9 | 2534 | 116 | |
XM_011531735.2 | XP_011530037.1 | 9 - 10 | 2690 | 116 | |
XM_017007878.1 | XP_016863367.1 | 8 - 9 | 2624 | 116 | |
XM_017007879.1 | XP_016863368.1 | 8 - 9 | 2621 | 116 | |
XM_017007880.1 | XP_016863369.1 | 7 - 8 | 2468 | 116 | |
XM_017007881.1 | XP_016863370.1 | 7 - 8 | 2465 | 116 | |
GenBank mRNA | AB209375.1 | - | 10 - 11 | 2683 | 116 |
AK307826.1 | - | 10 - 11 | 2122 | 116 | |
BC136257.1 | - | 10 - 11 | 1950 | 116 | |
BC136258.1 | - | 10 - 11 | 1950 | 116 | |
BC143427.1 | - | 9 - 10 | 1884 | 116 | |
BX537946.1 | - | 10 - 11 | 1802 | 116 | |
L36644.1 | - | 9 - 10 | 1789 | 116 |