Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EPHA5
Entrez Gene ID
2044
Gene Name
EPH receptor A5
Gene Aliases
CEK7, EHK-1, EHK1, EK7, HEK7, TYRO4
Chromosome Location
Chr.4: 65319563 - 65670838 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281765.2 NP_001268694.1 11 - 12 2806 147
NM_001281766.2 NP_001268695.1 10 - 11 2740 147
NM_001281767.2 NP_001268696.1 11 - 12 2806 147
NM_001318761.1 NP_001305690.1 11 - 12 1924 147
NM_004439.7 NP_004430.4 11 - 12 2803 147
NM_182472.4 NP_872272.2 10 - 11 2737 147
XM_005265653.3 XP_005265710.1 9 - 10 2657 147
XM_011531735.2 XP_011530037.1 10 - 11 2813 147
XM_017007878.1 XP_016863367.1 9 - 10 2747 147
XM_017007879.1 XP_016863368.1 9 - 10 2744 147
XM_017007880.1 XP_016863369.1 8 - 9 2591 147
XM_017007881.1 XP_016863370.1 8 - 9 2588 147
GenBank mRNA AB209375.1 - 11 - 12 2806 147
AK307826.1 - 11 - 12 2245 147
BC136257.1 - 11 - 12 2073 147
BC136258.1 - 11 - 12 2073 147
BC143427.1 - 10 - 11 2007 147
BX537946.1 - 11 - 12 1925 147
L36644.1 - 10 - 11 1912 147

View Full Product Details