The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001281765.2 | NP_001268694.1 | 1 - 2 | 931 | 94 |
NM_001281766.2 | NP_001268695.1 | 1 - 2 | 931 | 94 | |
NM_001281767.2 | NP_001268696.1 | 1 - 2 | 931 | 94 | |
NM_004439.7 | NP_004430.4 | 1 - 2 | 931 | 94 | |
NM_182472.4 | NP_872272.2 | 1 - 2 | 931 | 94 | |
XM_005265653.3 | XP_005265710.1 | 1 - 2 | 1274 | 94 | |
XM_011531735.2 | XP_011530037.1 | 1 - 2 | 1274 | 94 | |
XM_017007878.1 | XP_016863367.1 | 1 - 2 | 1274 | 94 | |
XM_017007879.1 | XP_016863368.1 | 1 - 2 | 1274 | 94 | |
XM_017007880.1 | XP_016863369.1 | 1 - 2 | 1274 | 94 | |
XM_017007881.1 | XP_016863370.1 | 1 - 2 | 1274 | 94 | |
GenBank mRNA | AK307826.1 | - | 1 - 2 | 371 | 94 |
BC136257.1 | - | 1 - 2 | 198 | 94 | |
BC143427.1 | - | 1 - 2 | 198 | 94 | |
L36644.1 | - | 1 - 2 | 106 | 94 |