The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098484.2 | NP_001091954.1 | 14 - 15 | 2022 | 73 |
NM_001134742.1 | NP_001128214.1 | 14 - 15 | 2022 | 73 | |
NM_003759.3 | NP_003750.1 | 11 - 12 | 1969 | 73 | |
XM_011532390.2 | XP_011530692.1 | 9 - 10 | 1508 | 73 | |
XM_017008792.1 | XP_016864281.1 | 12 - 13 | 1695 | 73 | |
XM_017008793.1 | XP_016864282.1 | 9 - 10 | 1994 | 73 | |
GenBank mRNA | AB470072.1 | - | 13 - 14 | 1912 | 73 |
AF007216.1 | - | 11 - 12 | 1922 | 73 | |
AF011390.1 | - | 14 - 15 | 2022 | 73 | |
AF053753.1 | - | 13 - 14 | 1905 | 73 | |
AF053754.1 | - | 13 - 14 | 1905 | 73 | |
AF069510.1 | - | 13 - 14 | 1949 | 73 | |
AF157492.1 | - | 14 - 15 | 1981 | 73 | |
AF310248.1 | - | 13 - 14 | 1905 | 73 | |
AK226164.1 | - | 12 - 13 | 1795 | 73 | |
AK307594.1 | - | 14 - 15 | 2002 | 73 | |
EF531618.1 | - | 13 - 14 | 1905 | 73 |