The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098484.2 | NP_001091954.1 | 9 - 10 | 1165 | 102 |
NM_001134742.1 | NP_001128214.1 | 9 - 10 | 1165 | 102 | |
NM_003759.3 | NP_003750.1 | 6 - 7 | 1112 | 102 | |
XM_011532390.2 | XP_011530692.1 | 4 - 5 | 651 | 102 | |
XM_017008792.1 | XP_016864281.1 | 7 - 8 | 838 | 102 | |
XM_017008793.1 | XP_016864282.1 | 4 - 5 | 1137 | 102 | |
GenBank mRNA | AB470072.1 | - | 8 - 9 | 1055 | 102 |
AF004813.1 | - | 7 - 8 | 988 | 102 | |
AF007216.1 | - | 6 - 7 | 1065 | 102 | |
AF011390.1 | - | 9 - 10 | 1165 | 102 | |
AF053753.1 | - | 8 - 9 | 1048 | 102 | |
AF069510.1 | - | 8 - 9 | 1092 | 102 | |
AF157492.1 | - | 9 - 10 | 1124 | 102 | |
AF310248.1 | - | 8 - 9 | 1048 | 102 | |
AK226164.1 | - | 7 - 8 | 938 | 102 | |
AK307594.1 | - | 9 - 10 | 1145 | 102 | |
AK308038.1 | - | 7 - 8 | 639 | 102 | |
BC030977.1 | - | 6 - 7 | 1112 | 102 | |
CR749482.1 | - | 1 - 2 | 70 | 102 | |
EF531618.1 | - | 8 - 9 | 1048 | 102 |