Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RBMS2
Entrez Gene ID
5939
Gene Name
RNA binding motif single stranded interacting protein 2
Gene Aliases
SCR3
Chromosome Location
Chr.12: 56521812 - 56596196 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002898.3 NP_002889.1 8 - 9 975 100
XM_005269059.4 XP_005269116.1 8 - 9 843 100
XM_005269060.4 XP_005269117.1 8 - 9 843 100
XM_005269061.3 XP_005269118.1 8 - 9 843 100
XM_006719541.3 XP_006719604.1 8 - 9 843 100
XM_006719542.3 XP_006719605.1 8 - 9 843 100
XM_006719543.3 XP_006719606.1 8 - 9 843 100
XM_006719544.3 XP_006719607.1 8 - 9 843 100
XM_011538637.2 XP_011536939.1 8 - 9 787 100
XM_011538638.2 XP_011536940.1 9 - 10 1109 100
XM_011538639.2 XP_011536941.1 8 - 9 843 100
XM_011538640.2 XP_011536942.1 8 - 9 843 100
XM_011538642.2 XP_011536944.1 6 - 7 573 100
XM_017019775.1 XP_016875264.1 8 - 9 782 100
XM_017019776.1 XP_016875265.1 4 - 5 267 100
XM_017019777.1 XP_016875266.1 7 - 8 691 100
XM_017019778.1 XP_016875267.1 4 - 5 283 100
GenBank mRNA AK293315.1 - 7 - 8 575 100
AK304540.1 - 7 - 8 635 100
AK308885.1 - 6 - 7 671 100
BC027863.1 - 8 - 9 784 100
BC072679.1 - 8 - 9 848 100
D28483.1 - 8 - 9 792 100

View Full Product Details