The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000885.5 | NP_000876.3 | 19 - 20 | 2934 | 87 |
GenBank mRNA | AB208885.1 | - | 16 - 17 | 1712 | 87 |
AK310610.1 | - | 5 - 6 | 682 | 87 | |
BC080190.1 | - | 2 - 3 | 279 | 87 | |
L12002.1 | - | 19 - 20 | 2599 | 87 | |
X16983.1 | - | 19 - 20 | 2213 | 87 |