The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001835.3 | NP_001826.3 | 18 - 19 | 2994 | 65 |
NM_007098.3 | NP_009029.3 | 18 - 19 | 2994 | 65 | |
XM_011530401.2 | XP_011528703.1 | 8 - 9 | 1228 | 65 | |
XM_017028953.1 | XP_016884442.1 | 18 - 19 | 2987 | 65 | |
XM_017028954.1 | XP_016884443.1 | 17 - 18 | 2873 | 65 | |
XM_017028955.1 | XP_016884444.1 | 18 - 19 | 2987 | 65 | |
XM_017028956.1 | XP_016884445.1 | 18 - 19 | 2987 | 65 | |
XM_017028957.1 | XP_016884446.1 | 18 - 19 | 2987 | 65 | |
GenBank mRNA | AK302506.1 | - | 18 - 19 | 2963 | 65 |
BC143977.1 | - | 18 - 19 | 2939 | 65 | |
BC143979.1 | - | 18 - 19 | 2939 | 65 | |
L77568.1 | - | 4 - 5 | 627 | 65 | |
U41763.1 | - | 18 - 19 | 3016 | 65 | |
U60802.1 | - | 18 - 19 | 2963 | 65 | |
X95486.1 | - | 18 - 19 | 2952 | 65 |