The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001835.3 | NP_001826.3 | 31 - 32 | 4853 | 84 |
NM_007098.3 | NP_009029.3 | 32 - 33 | 5024 | 84 | |
XM_011530401.2 | XP_011528703.1 | 22 - 23 | 3258 | 84 | |
XM_017028953.1 | XP_016884442.1 | 32 - 33 | 4993 | 84 | |
XM_017028954.1 | XP_016884443.1 | 31 - 32 | 4903 | 84 | |
XM_017028955.1 | XP_016884444.1 | 31 - 32 | 4801 | 84 | |
XM_017028956.1 | XP_016884445.1 | 30 - 31 | 4735 | 84 | |
GenBank mRNA | AK125929.1 | - | 2 - 3 | 4147 | 84 |
AK294147.1 | - | 9 - 10 | 1226 | 84 | |
AK295264.1 | - | 11 - 12 | 1506 | 84 | |
AK302506.1 | - | 31 - 32 | 4822 | 84 | |
L77568.1 | - | 18 - 19 | 2536 | 84 | |
U41763.1 | - | 32 - 33 | 5046 | 84 | |
U60802.1 | - | 32 - 33 | 4995 | 84 | |
X95486.1 | - | 32 - 33 | 4982 | 84 | |
X95487.1 | - | 4 - 5 | 430 | 84 |