Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PNRC2
Entrez Gene ID
55629
Gene Name
proline rich nuclear receptor coactivator 2
Gene Aliases
-
Chromosome Location
Chr.1: 23959164 - 23963462 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017001691.1 XP_016857180.1 1 - 2 89 78

View Full Product Details