The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017446.3 | NP_059142.2 | 6 - 7 | 713 | 99 |
NM_080794.3 | NP_542984.2 | 6 - 7 | 713 | 99 | |
XM_006724026.3 | XP_006724089.1 | 6 - 7 | 705 | 99 | |
XM_011529651.2 | XP_011527953.1 | 6 - 7 | 784 | 99 | |
GenBank mRNA | AF109357.1 | - | 6 - 7 | 687 | 99 |
AF239727.1 | - | 6 - 7 | 699 | 99 | |
AF270511.1 | - | 3 - 4 | 252 | 99 | |
AK000458.1 | - | 6 - 7 | 704 | 99 | |
AK225878.1 | - | 6 - 7 | 704 | 99 | |
BC004896.2 | - | 6 - 7 | 686 | 99 | |
BC107719.1 | - | 6 - 7 | 709 | 99 | |
CD368044.1 | - | 4 - 5 | 99 |