The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032139.2 | NP_115515.2 | 14 - 15 | 1488 | 64 |
XM_006723412.2 | XP_006723475.1 | 14 - 15 | 1381 | 64 | |
XM_017027349.1 | XP_016882838.1 | 13 - 14 | 1350 | 64 | |
GenBank mRNA | AK054561.1 | - | 14 - 15 | 1477 | 64 |
AL136784.1 | - | 14 - 15 | 1465 | 64 | |
BC045605.1 | - | 14 - 15 | 1493 | 64 | |
BC050529.1 | - | 14 - 15 | 1444 | 64 |