The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032139.2 | NP_115515.2 | 2 - 3 | 263 | 60 |
XM_006723412.2 | XP_006723475.1 | 2 - 3 | 156 | 60 | |
XM_017027349.1 | XP_016882838.1 | 2 - 3 | 282 | 60 | |
GenBank mRNA | AF447882.1 | - | 2 - 3 | 237 | 60 |
AK054561.1 | - | 2 - 3 | 252 | 60 | |
AL136784.1 | - | 2 - 3 | 240 | 60 | |
BC045605.1 | - | 2 - 3 | 268 | 60 | |
BC050529.1 | - | 2 - 3 | 219 | 60 | |
BI462844.1 | - | - | 272 | 60 |