The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001315.2 | NP_001306.1 | 10 - 11 | 1327 | 104 |
NM_139012.2 | NP_620581.1 | 10 - 11 | 1327 | 104 | |
XM_006714998.3 | XP_006715061.1 | 10 - 11 | 802 | 104 | |
XM_011514310.2 | XP_011512612.2 | 10 - 11 | 1595 | 104 | |
GenBank mRNA | AF100544.1 | - | 10 - 11 | 861 | 104 |
AK291709.1 | - | 10 - 11 | 1222 | 104 | |
AK303414.1 | - | 11 - 12 | 891 | 104 | |
BC000092.1 | - | 10 - 11 | 1184 | 104 | |
BC031574.1 | - | 10 - 11 | 1183 | 104 | |
BT006933.1 | - | 10 - 11 | 846 | 104 | |
CR536505.1 | - | 10 - 11 | 846 | 104 | |
L35263.1 | - | 10 - 11 | 1208 | 104 | |
L35264.1 | - | 10 - 11 | 1054 | 104 |