The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017434.4 | NP_059130.2 | 12 - 13 | 1563 | 73 |
NM_175940.2 | NP_787954.1 | 11 - 12 | 1371 | 73 | |
XM_011521681.2 | XP_011519983.1 | 10 - 11 | 1264 | 73 | |
GenBank mRNA | AF213465.1 | - | 11 - 12 | 1371 | 73 |
AF230495.1 | - | 12 - 13 | 1563 | 73 | |
AK075268.1 | - | 11 - 12 | 1372 | 73 | |
AK290372.1 | - | 12 - 13 | 1624 | 73 | |
BC114628.1 | - | 11 - 12 | 1360 | 73 | |
BC114938.1 | - | 11 - 12 | 1360 | 73 |