The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017434.4 | NP_059130.2 | 19 - 20 | 2661 | 65 |
NM_175940.2 | NP_787954.1 | 18 - 19 | 2469 | 65 | |
XM_011521681.2 | XP_011519983.1 | 17 - 18 | 2362 | 65 | |
XM_011521682.1 | XP_011519984.1 | 8 - 9 | 1272 | 65 | |
GenBank mRNA | AF213465.1 | - | 18 - 19 | 2469 | 65 |
AF230495.1 | - | 19 - 20 | 2661 | 65 | |
AK075268.1 | - | 18 - 19 | 2470 | 65 | |
AK128591.1 | - | 8 - 9 | 1526 | 65 | |
AK290372.1 | - | 19 - 20 | 2722 | 65 | |
BC114628.1 | - | 18 - 19 | 2458 | 65 | |
BC114938.1 | - | 18 - 19 | 2458 | 65 |