The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014044.5 | NP_054763.2 | 2 - 3 | 430 | 68 |
XM_005263913.4 | XP_005263970.1 | 2 - 3 | 526 | 68 | |
XM_006712403.3 | XP_006712466.1 | 2 - 3 | 526 | 68 | |
XM_006712404.2 | XP_006712467.1 | 2 - 3 | 440 | 68 | |
GenBank mRNA | AF077038.1 | - | 2 - 3 | 382 | 68 |
AK222599.1 | - | 2 - 3 | 396 | 68 | |
AL080115.1 | - | 2 - 3 | 434 | 68 | |
AY017215.1 | - | 2 - 3 | 398 | 68 | |
AY251534.1 | - | 1 - 2 | 291 | 68 | |
BC017019.1 | - | 2 - 3 | 563 | 68 | |
BC025992.2 | - | 1 - 2 | 1458 | 68 | |
BC039078.1 | - | 1 - 2 | 1434 | 68 | |
BC091481.1 | - | 2 - 3 | 420 | 68 | |
CR533464.1 | - | 1 - 2 | 278 | 68 | |
KU178586.1 | - | 1 - 2 | 278 | 68 | |
KU178587.1 | - | 1 - 2 | 278 | 68 |