Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACAN
Entrez Gene ID
176
Gene Name
aggrecan
Gene Aliases
AGC1, AGCAN, CSPG1, CSPGCP, MSK16, SEDK
Chromosome Location
Chr.15: 88803442 - 88875354 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013227.3 NP_037359.3 12 - 13 7205 100
XM_006720419.1 XP_006720482.1 12 - 13 7206 100
XM_017021985.1 XP_016877474.1 12 - 13 7206 100
GenBank mRNA J05062.1 - 1 - 2 681 100

View Full Product Details