Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACAN
Entrez Gene ID
176
Gene Name
aggrecan
Gene Aliases
AGC1, AGCAN, CSPG1, CSPGCP, MSK16, SEDK
Chromosome Location
Chr.15: 88803442 - 88875354 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135.3 NP_001126.3 3 - 4 827 107
NM_013227.3 NP_037359.3 3 - 4 827 107
XM_006720419.1 XP_006720482.1 3 - 4 828 107
XM_011521313.1 XP_011519615.1 3 - 4 828 107
XM_011521314.1 XP_011519616.1 3 - 4 828 107
XM_017021985.1 XP_016877474.1 3 - 4 828 107
XM_017021986.1 XP_016877475.1 3 - 4 828 107
XM_017021987.1 XP_016877476.1 3 - 4 828 107
GenBank mRNA AK290723.1 - 3 - 4 827 107
BC036445.1 - 3 - 4 798 107
BC150624.1 - - 511 107
M55172.1 - 3 - 4 513 107

View Full Product Details