The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022167.3 | NP_071450.2 | 7 - 8 | 1597 | 85 |
NR_110010.1 | 7 - 8 | 1597 | 85 | ||
XM_005257572.4 | XP_005257629.1 | 7 - 8 | 1911 | 85 | |
GenBank mRNA | AJ277442.1 | - | 7 - 8 | 1597 | 85 |
AK299999.1 | - | 7 - 8 | 1494 | 85 | |
AY358090.1 | - | 7 - 8 | 1493 | 85 | |
BC052262.1 | - | 7 - 8 | 1517 | 85 |