The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012138.3 | NP_036270.1 | 11 - 12 | 1868 | 90 |
GenBank mRNA | AF083208.1 | - | 11 - 12 | 1785 | 90 |
AF161395.1 | - | 6 - 7 | 538 | 90 | |
AJ249940.2 | - | 11 - 12 | 1723 | 90 | |
AK026173.1 | - | 9 - 10 | 1294 | 90 | |
AK057229.1 | - | 11 - 12 | 1868 | 90 | |
BC000591.2 | - | 11 - 12 | 1748 | 90 |