Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GGNBP2
Entrez Gene ID
79893
Gene Name
gametogenetin binding protein 2
Gene Aliases
DIF-3, DIF3, LCRG1, LZK1, ZFP403, ZNF403
Chromosome Location
Chr.17: 36544888 - 36589848 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024835.4 NP_079111.1 13 - 14 2204 146
XM_005257689.2 XP_005257746.1 13 - 14 2186 146
XM_011525265.2 XP_011523567.1 12 - 13 1949 146
XM_011525266.1 XP_011523568.1 12 - 13 2072 146
XM_017025107.1 XP_016880596.1 12 - 13 1917 146
GenBank mRNA AF126964.1 - 12 - 13 1935 146
AJ404670.2 - 5 - 6 761 146
AK026214.1 - 5 - 6 838 146
AK290687.1 - 12 - 13 1960 146
AK301614.1 - 10 - 11 1492 146
BC137491.1 - 13 - 14 2070 146

View Full Product Details