Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GGNBP2
Entrez Gene ID
79893
Gene Name
gametogenetin binding protein 2
Gene Aliases
DIF-3, DIF3, LCRG1, LZK1, ZFP403, ZNF403
Chromosome Location
Chr.17: 36544888 - 36589848 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024835.4 NP_079111.1 5 - 6 843 67
XM_005257689.2 XP_005257746.1 5 - 6 819 67
XM_011525265.2 XP_011523567.1 4 - 5 582 67
XM_017025107.1 XP_016880596.1 4 - 5 556 67
GenBank mRNA AF126964.1 - 4 - 5 574 67
AF268387.1 - 5 - 6 763 67
AK024883.1 - 5 - 6 799 67
AK290687.1 - 4 - 5 599 67
AK292161.1 - 5 - 6 812 67
AK301614.1 - 2 - 3 131 67
BC137491.1 - 5 - 6 709 67

View Full Product Details