Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRPSAP1
Entrez Gene ID
5635
Gene Name
phosphoribosyl pyrophosphate synthetase associated protein 1
Gene Aliases
PAP39
Chromosome Location
Chr.17: 76310787 - 76354149 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002766.2 NP_002757.2 9 - 10 1445 68
XM_011525034.2 XP_011523336.1 8 - 9 1143 68
GenBank mRNA AK298171.1 - 9 - 10 911 68
AK312637.1 - 9 - 10 1156 68
BC009012.1 - 9 - 10 924 68
D61391.1 - 10 - 11 961 68

View Full Product Details