Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CA5B
Entrez Gene ID
11238
Gene Name
carbonic anhydrase 5B
Gene Aliases
CA-VB
Chromosome Location
Chr.X: 15738281 - 15787625 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005274442.4 XP_005274499.1 2 - 3 142 117

View Full Product Details