The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001169574.1 | NP_001163045.1 | 7 - 8 | 1128 | 65 |
NM_152761.2 | NP_689974.2 | 7 - 8 | 1128 | 65 | |
XM_011545473.2 | XP_011543775.1 | 7 - 8 | 885 | 65 | |
XM_011545474.2 | XP_011543776.1 | 3 - 4 | 418 | 65 | |
GenBank mRNA | AK058173.1 | - | 7 - 8 | 1128 | 65 |
BC117344.1 | - | 7 - 8 | 1040 | 65 | |
BC117346.1 | - | 7 - 8 | 1040 | 65 | |
BC144007.1 | - | 7 - 8 | 1040 | 65 |